Learn More
Thermo Scientific™ GC SMART Syringes for TriPlus™ RSH SMART Autosampler
Product Code. 17617065 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
Needle Gauge:
19 G
22 G
23 G
23s G
26 G
26s G
Length (Metric) Needle:
57 mm
57 or 85 mm
65 mm
85 mm
Product Type:
GC Autosampler Syringe, Bevel
GC Autosampler Syringe, Cone
Gas-tight GC Autosampler Syringe, Cone
Gas-tight GC Autosampler Syringe, LC
Gas-tight GC Autosampler Syringe, Side Hole
Removable Needle Syringe, needle not included
Unit Size:
Each
39 product options available for selection
Product selection table with 39 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Length (Metric) Needle Product Type Unit Size
17617065 10 μL 26s G 57 mm GC Autosampler Syringe, Cone Each
17627055 0.5 μL 23 G 57 mm GC Autosampler Syringe, Cone Each
17637055 1 μL 23 G 57 mm GC Autosampler Syringe, Cone Each
17647055 5 μL 23s G 85 mm GC Autosampler Syringe, Cone Each
17657055 5 μL 23s G 57 mm GC Autosampler Syringe, Cone Each
17667055 5 μL 26s G 85 mm GC Autosampler Syringe, Cone Each
17677055 5 μL 26s G 57 mm GC Autosampler Syringe, Cone Each
17687055 10 μL 23s G 85 mm GC Autosampler Syringe, Cone Each
17697055 10 μL 23s G 57 mm GC Autosampler Syringe, Cone Each
17607065 10 μL 26s G 85 mm GC Autosampler Syringe, Cone Each
17627065 10 μL 23s G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17637065 10 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17647065 10 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17657065 10 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17667065 10 μL 26s G 57 mm GC Autosampler Syringe, Bevel Each
17677065 25 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17687065 25 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17697065 25 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17607075 50 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17617075 50 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17627075 50 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17637075 100 μL 23s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17647075 100 μL 26s G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17657075 100 μL 26s G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17667075 100 μL 23 G 85 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17677075 100 μL 23 G 57 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17687075 250 μL 26 G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17697075 250 μL 26 G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17607085 250 μL 23 G 85 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17617085 250 μL 23 G 57 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17627085 500 μL 26 G 85 mm Gas-tight GC Autosampler Syringe, Cone Each
17637085 500 μL 26 G 57 mm Gas-tight GC Autosampler Syringe, Cone Each
17647085 500 μL 23 G 57 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17657085 1 mL 22 G 57 mm Gas-tight GC Autosampler Syringe, LC Each
17667085 1 mL 23 G 65 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17677085 2.5 mL 23 G 65 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17687085 5 mL 23 G 65 mm Gas-tight GC Autosampler Syringe, Side Hole Each
17697085 10 mL 19 G 57 mm Gas-tight GC Autosampler Syringe, LC Each
31034046 2.5 mL 22 G 57 or 85 mm Removable Needle Syringe, needle not included Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
39 options
Product Code. 17617065 Supplier Thermo Scientific™ Supplier No. 365D0291SM
Save Up to 60% on Contract Testing Essentials
Use promo code 28025
View all promotions
In Stock
Add to basket
Edge
Add to basket
Use these GC syringes embedded with an ID chip to track part and lot numbers, usage, and history to increase data confidence and ensure system compliance.
Use the unique technology of Thermo Scientific™ GC SMART Syringes as a traceable usage-based approach to gas chromatography (GC) syringe management, resulting in increased reliability, instrument up-time, confidence in the results, and full traceability. When installed on a Thermo Scientific™ TriPlus™ RSH SMART autosampler, the SMART syringes benefit from the unique SMART technology. Each syringe is equipped with an ID chip, which stores information such as part number, lot number, usage range, and history for that specific syringe. The information stored on the chip can be directly accessed and reported via Thermo Scientific™ Chromeleon™ CDS software.
SMART syringes are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.
SMART Technology Benefits
- Gain higher confidence in analytical results
- Ensure error free usage of syringes through automatic identification
- Support GLP accreditation with usage history traceability
- Optimize consumption by monitoring of usage conditions and lifetime
- Facilitate syringe management thanks to color-coded health notification dashboard integrated in Chromeleon™ CDS
TRUSTED_SUSTAINABILITY
Specifications
Description 10 μL FN, 26 s G, Cone, 57 mm
Needle Gauge 26s G
Product Type GC Autosampler Syringe, Cone
Volume (Metric) 10 μL
Length (Metric) Needle 57 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus RSH SMART Autosampler, All TriPlus RSH Instruments and All PAL3 Models