Learn More
Trajan Scientific and Medical Gas Filter Connecting Unit
The gas filter connecting unit has inlet and outlet connectors for the gas line and the system can be wall-mounted or fixed to a bench. Connecting units come with a 1, 2 or 4 filter capacity and are available for 1/4 in. or 1/8 in. gas lines
283.00€ - 1030.00€
Specifications
For Use With (Equipment) Gas filters
Packaging Quantity Each
View More Specs
Products 6
Product Code Brand Thread Size Type Price Quantity & Availability  
Product Code Brand Thread Size Type Price Quantity & Availability  
15750959
View Documents Promotion Details
Trajan Scientific and Medical
1035018
1/8 in. 1-position Connecting Unit
283.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
15740959
View Documents Promotion Details
Trajan Scientific and Medical
1035014
1/4 in. 1-position Connecting Unit
283.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
15770959
View Documents Promotion Details
Trajan Scientific and Medical
1035028
1/8 in. 2-position Connecting Unit
540.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
15760959
View Documents Promotion Details
Trajan Scientific and Medical
1035024
1/4 in. 2-position Connecting Unit
540.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
15790959
View Documents Promotion Details
Trajan Scientific and Medical
1035048
1/8 in. 4-position Connecting Unit
1030.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
15780959
View Documents Promotion Details
Trajan Scientific and Medical
1035044
1/4 in. 4-position Connecting Unit
1030.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
Description
  • Easily installed into the gas line
  • Can be either wall-mounted or fixed to a bench
  • Available with a 1, 2 or 4 filter capacity
Specifications
Specifications
Gas filters