Learn More
ROBU VitraPOR™ Gas Distribution Tube, Type A
Gas Distribution Tube with centered bowl.
178.00€ - 238.00€
Product Code | Brand | Pore Size | Diameter (Metric) Inner Tubing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) Inner Tubing | Price | Quantity & Availability | ||||
16812986
|
ROBU VitraPOR™
19251 |
1 | 25 mm |
178.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16802986
|
ROBU VitraPOR™
19250 |
0 | 25 mm |
178.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16852986
|
ROBU VitraPOR™
19340 |
0 | 34 mm |
203.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16842986
|
ROBU VitraPOR™
19254 |
4 | 25 mm |
209.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16872986
|
ROBU VitraPOR™
19342 |
2 | 34 mm |
203.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16882986
|
ROBU VitraPOR™
19343 |
3 | 34 mm |
203.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16892986
|
ROBU VitraPOR™
19344 |
4 | 34 mm |
238.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16832986
|
ROBU VitraPOR™
19253 |
3 | 25 mm |
178.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16862986
|
ROBU VitraPOR™
19341 |
1 | 34 mm |
203.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16822986
|
ROBU VitraPOR™
19252 |
2 | 25 mm |
178.00€
Pack of 5 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
Description
- For fine dispersion of gases in liquids
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Type A |