Learn More
Brady™ Gas Cylinder, 3 Part Tag
Description
- Add the Cylinder ID No. to each section and as it goes through it's life cycle, tear off the sections (Full, In Use, Empty)
- Single Sided tag with whole punch to allow cable tie attachment
- Space for logo, location or contractor information
- Can be customized
- Utilize with fine nibbed permanent marker pen for durability
- Ultra tear-resistant and have been proven to withstand some of the harshest environments
- 100% Recyclable
Specifications
Specifications
| Alternate Part Number | PROC-GB-PT 001-50 |
| Color | Black, Red, Yellow, Green, White |
| Material | Polypropylene |
| Height (Metric) | 150 mm |
| Width (Metric) | 80 mm |