Learn More
MP Biomedicals™ G418 Disulfate PhytoPure™
G418 Disulfate PhytoPure™ is an aminoglycoside related to gentamicin that acts as an antibiotic and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths.
Brand: MP Biomedicals™ 0215878291
301.69 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : CAS : 1405-41-0 Weight : 1.36080kg
Description
- G418 Disulfate blocks polypeptide synthesis in both prokaryotic and eukaryotic cells and is useful for selection of genetically engineered eukarytoic cells in culture.
- It is an effective selecting agent utilized in killing prokaryotic and eukaryotic cells.
- It is also used for selection and maintenance of prokaryotic and eukaryotic cells transfected with a iNOS promoter construct and neomycin resistance gene.
Specifications
108321-42-2 | |
The powder should be stored away from light and moisture. Solutions should be aliquoted and stored frozen (− 5 to − 20°C). | |
Antibiotic | |
Cell Culture Grade |
White | |
Powder | |
Non-sterile | |
1 g |
Safety and Handling
- Geneticin Disulfate Salt, cell culture grade
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P321-Specific treatment (see on this label).
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Geneticin DisulfateSalt
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.