Learn More
Fungilab™ Stainless Steel Spindles
Accurately measure viscosities with these stainless steel spindles. Stainless Steel Spindles come in a variety of volumes and viscosity ranges.
POA
Specifications
For Use With (Equipment) | Small Sample Adapter |
---|---|
Certifications/Compliance | ASTM, ISO 2555 |
Material | Stainless Steel |
For Use With (Application) | For relative viscosity measurement |
Description
- Spindles for use with Small Sample Adapters
- Used for measurements in accordance with the ASTM-ISO 2555 standard
- Small sample adapter works with special spindes TR for R and H viscometer type and TL for L viscometer type
Specifications
Small Sample Adapter | |
ASTM, ISO 2555 | |
Stainless Steel | |
For relative viscosity measurement |