Learn More
Fungilab™ Stainless Steel Spindles
Accurately measure viscosities with these stainless steel spindles. Stainless Steel Spindles come in a variety of volumes and viscosity ranges.
POA
Specifications
For Use With (Equipment) Small Sample Adapter
Certifications/Compliance ASTM, ISO 2555
Material Stainless Steel
For Use With (Application) For relative viscosity measurement
View More Specs
Description
Description
  • Spindles for use with Small Sample Adapters
  • Used for measurements in accordance with the ASTM-ISO 2555 standard
  • Small sample adapter works with special spindes TR for R and H viscometer type and TL for L viscometer type
Specifications
Specifications
Small Sample Adapter
ASTM, ISO 2555
Stainless Steel
For relative viscosity measurement