Learn More
Labconco FreeZone 12L Console Freeze Dryer
FreeZone Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. The console features a front viewing window, interior space for a vacuum pump, and accommodation for a large tray dryer.
28121.45€ - 67622.02€
Specifications
Type | Console Freeze Dryer |
---|---|
Capacity (Metric) | 12 L |
Plug Type | All except North America |
Voltage | 230 V |
Product Code | Brand | Material | Temperature Range (Metric) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Temperature Range (Metric) | Includes | Price | Quantity & Availability | |||
17921040
|
Labconco
701221090 |
Stainless Steel Coil and Collector Chamber | -50°C | Stoppering Tray Dryer |
59752.18€
1 unit |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
17971040
|
Labconco
701221190 |
Stainless Steel Coil and Collector Chamber | -50°C | Purge Valve |
61369.66€
1 unit |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
17981040
|
Labconco
701222090 |
PTFE Coil and Collector Chamber | -50°C | Stoppering Tray Dryer |
61459.52€
1 unit |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
17951050
|
Labconco
701222190 |
PTFE Coil and Collector Chamber | -50°C | Purge Valve, Stoppering Tray Dryer |
63062.81€
1 unit |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
17900180
|
Labconco
711211090 |
Stainless Steel Coil and Collector Chamber | -84°C |
28121.45€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
17950150
|
Labconco
711211190 |
Stainless Steel Coil and Collector Chamber | -84°C | Purge Valve |
29729.47€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17990170
|
Labconco
711211290 |
Stainless Steel Coil and Collector Chamber | -84°C | Purge Valve, Mini Chamber |
32496.22€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17910160
|
Labconco
711212090 |
PTFE Coil and Collector Chamber | -84°C |
29833.52€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
17980170
|
Labconco
711212190 |
PTFE Coil and Collector Chamber | -84°C | Purge Valve |
31446.27€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17910140
|
Labconco
711212290 |
PTFE Coil and Collector Chamber | -84°C | Purge Valve, Mini Chamber |
34208.29€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17971050
|
Labconco
711221090 |
Stainless Steel Coil and Collector Chamber | -84°C | Stoppering Tray Dryer |
64297.20€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17901040
|
Labconco
711221190 |
Stainless Steel Coil and Collector Chamber | -84°C | Purge Valve, Stoppering Tray Dryer |
65905.22€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17961040
|
Labconco
711222090 |
PTFE Coil and Collector Chamber | -84°C | Stoppering Tray Dryer |
66009.27€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
17981050
|
Labconco
711222190 |
PTFE Coil and Collector Chamber | -84°C | Purge Valve, Stoppering Tray Dryer |
67622.02€
1 unit |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
Description
- Coil holds 12 liters of ice before defrosting
- The hp HCFC/CFC-free refrigeration system cools the collector to -50° C (-58° F), suitable for aqueous samples.
- Includes a stoppering tray dryer with heated and cooled shelves, vacuum release valve control, stoppering knob and more.
- Lyo-Works™ OS provides a real time display of collector temperature, vacuum level and optional End-Zone™ end point detection system
- Large, intuitive 5 in. full color, capacitive touchscreen display for easy operation.
- Stay Connected
- View Real-Time Sample Data
- Get Consistent, High Quality Results
- Make Challenging Samples Easy
Specifications
Console Freeze Dryer | |
12 L | |
All except North America | |
230 V |