Learn More
Thermo Scientific™ Freezer and Refrigerator Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Brand: Thermo Scientific™ 7125
Additional Details : Weight : 0.01000kg
Description
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
- Epoxy-coated open-wire shelves
- Full- and half-depth sizes
- Undercounter units use full-depth shelves only
Specifications
Epoxy-coated open wire shelves | |
11.5 cu. ft. | |
Epoxy Coated Steel | |
4.7 cu. ft. (133L), 11.5 cu. ft. (326L) units | |
General Freezer Items |
Epoxy-Coated Open Wire Shelves | |
326 L | |
Plastic | |
Warranty varies by country, please contact us for details |