Learn More
Esculape Laboratoire™ Portable First Aid Kit in Red Pouch
Product Code. 11718383
Change view
Click to view available options
Width (Metric):
50 mm
170 mm
Sufficient For:
1 to 4 People
4 to 10 People
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Sufficient For Unit Size
11718383 170 mm 4 to 10 People Each
11778383 50 mm 1 to 4 People Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11718383 Supplier Esculape Laboratoire™ Supplier No. 718 01 05
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
Handle emergency medical treatment with the Esculape™ Portable First Aid Kit in Red Pouch . It includes the essential tools, bandages and burn components to handle immediate care.
  • Hydrogel burn compress helps to reduce swelling While providing immediate cooling, relieves pain and moisturizes the skin.
  • Survival blanket
  • Bandages, compresses, forceps. gloves
Specifications
Depth (Metric) 170 mm
For Use With (Application) In all workplaces
Length (Metric) 230 mm
Product Type First Aid Kit
Color Red
Includes 5 Sterile compresses 200 x 200mm, 1 Gauze bandage 70 x 3000mm, 1 expandable tape 3000 x 70mm, 1 Sterile pressure dressing 2500 m x 60mm, 1 Sterile pressure dressing 100 x 2500mm, 10 assorted adhesive bandages, 1 Tape roll, 2 Alcoholic compresses, 2 Mosquito gauze, Anti-compress 2 strokes, 2 scald gel sachets, 2 Bactericidal soap bags, Scissors 1 pair, Splinter forceps 1, Clips 4, Vinyl gloves 1 pair, 1 Guide
Sufficient For 4 to 10 People
Width (Metric) 170 mm
Height (Metric) 50 mm