Learn More
Glassco Laboratory Equipments Flat Bottom Glass Flask
Description
- Flat bottom glass flasks with an NS 29/32 joint
- Available in 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL capacities
- Made from 3.3 borosilicate glass
- Complies with DIN/ISO 4797 and USP standards
- Features white printing
- Chemical resistant
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass (3.3) |
| Product Type | Flask |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Mouth Style | Joint |
| Sterility | Non-sterile |
| Bottom Shape | Flat |