Learn More
Labconco™ Flask Adapter
Product Code. 11955502 Shop All Labconco Corporation Products
Change view
Click to view available options
Diameter (English):
0.75 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Unit Size
11955502 0.75 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11955502 Supplier Labconco™ Supplier No. 7543800
  • ONLINE EXCLUSIVE PRICE
    76.95€
    62.68€ / Pack of 1
    Save 14.27€
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
Suitable adapter for connecting 29/32 round-bottom flasks to valve ports on drying chambers and manifolds. Labconco™ Flask Adapter is constructed of stainless steel.
  • Stainless steel construction
  • Suitable for use with freeze dryers
Specifications
Material Stainless Steel
Standard Taper Joint 29/32
For Use With (Equipment) Freeze dryers
Diameter (English) 0.75 in.
Dimensions (L x W) 3.5 in. wide, 0.75 in. diameter
Weight (Metric) 0.5 kg