Learn More
Fisherbrand™ Sterile Sampling Bags with Flat-Wire Closures
Product Code. 17903182 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
10 x 10 in.
14 x 10 in. (35.6 x 25.4cm)
15 x 5.5 in. (38.1 x 14cm)
9 x 4.5 in. (22.9 x 11.4cm)
9 x 5.5 in. (22.9 x 14cm)
Thickness (English):
3 mil
3.50 mil
4 mil
Capacity (English):
18 oz.
24 oz.
42 oz.
60 oz.
100 oz.
Color:
Clear
Clear with White Block
Unit Size:
Pack of 250
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Thickness (English) Capacity (English) Color Unit Size
17903182 9 x 4.5 in. (22.9 x 11.4cm) 3 mil 18 oz. Clear Pack of 500
13158167 9 x 5.5 in. (22.9 x 14cm) 3.50 mil 24 oz. Clear Pack of 500
11904405 9 x 5.5 in. (22.9 x 14cm) 3.50 mil 24 oz. Clear with White Block Pack of 500
11914405 10 x 10 in. 4 mil 60 oz. Clear with White Block Pack of 250
11738046 14 x 10 in. (35.6 x 25.4cm) 4 mil 100 oz. Clear Pack of 250
11974405 15 x 5.5 in. (38.1 x 14cm) 3.50 mil 42 oz. Clear Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17903182
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
An economical and efficient way to collect, contain and carry samples
Collect, contain and carry samples in an economical and efficient way
  • Sterile to avoid sample contamination
Specifications
Closure Type Flat Wire
Color Clear
Material Polyethylene
Thickness (English) 3 mil
Dimensions (L x W) 9 x 4.5 in. (22.9 x 11.4cm)
Length (English) 9 in.
Width (English) 4.5 in.
Capacity (English) 18 oz.
Length (Metric) 22.9 cm
Width (Metric) 11.4 cm