Learn More
Fisherbrand™ Sterile Sampling Bags with Flat-Wire Closures
Product Code. 11914405 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
10 x 10 in.
14 x 10 in. (35.6 x 25.4cm)
9 x 4.5 in. (22.9 x 11.4cm)
9 x 5.5 in. (22.9 x 14cm)
Thickness (English):
3 mil
3.50 mil
4 mil
Capacity (English):
18 oz.
24 oz.
60 oz.
100 oz.
Color:
Clear
Clear with White Block
Unit Size:
Pack of 250
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Thickness (English) Capacity (English) Color Unit Size
11914405 10 x 10 in. 4 mil 60 oz. Clear with White Block Pack of 250
17903182 9 x 4.5 in. (22.9 x 11.4cm) 3 mil 18 oz. Clear Pack of 500
13158167 9 x 5.5 in. (22.9 x 14cm) 3.50 mil 24 oz. Clear Pack of 500
11904405 9 x 5.5 in. (22.9 x 14cm) 3.50 mil 24 oz. Clear with White Block Pack of 500
11738046 14 x 10 in. (35.6 x 25.4cm) 4 mil 100 oz. Clear Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11914405
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket
An economical and efficient way to collect, contain and carry samples
Collect, contain and carry samples in an economical and efficient way
  • Sterile to avoid sample contamination
Specifications
Color Clear with White Block
Material Polyethylene
Sterility Sterile
Quantity 250/Pk.
Thickness (English) 4 mil
Dimensions (L x W) 10 x 10 in.
Length (English) 10 in.
Width (English) 10 in.
Product Type Sample Bag
Capacity (English) 60 oz.
Length (Metric) 25.4 cm
Width (Metric) 25.4 cm