Learn More
Fisherbrand™ Standard Rotor for High Speed Midi-Centrifuge
GREENER_CHOICE
Product Code. 12902051 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity:
1.5 to 2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
12902051 1.5 to 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12902051
  • ONLINE EXCLUSIVE PRICE
    75.20€
    56.75€ / Pack of 1
    Save 18.45€
    24% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Standard Rotor for High-Speed Midi-Centrifuge.
  • Standard Rotor for use with Fisherbrand™ High-Speed Midi-Centrifuge
  • Black
Specifications
Material Polycarbonate
Type Standard Rotor
Capacity 1.5 to 2 mL
Max. RCF None
Max. Speed None
For Use With (Equipment) Gusto™ High Speed Midi Centrifuge
No. of Places 12
Biocontainment No