Learn More
Fisherbrand™ Standard Pipette Tips
Standard tips for pipetters up to 1000μL
Description
For use with, but not limited to:
- Costar™ Onepette™, Costar 8/12 Pette™
- Eppendorf™
- Thermo Scientific Finnpipette
- LabPette™ 8- and 12-Channel
- Nichiryo™
- Pipetman™
- Rainin™ EDP2™
- Oxford™ Benchmate™
- Socorex™
- Titertek Digital™
- UNITEK™ Volac™
Specifications
| Universal |