Learn More
Fisherbrand™ Clean-Wipes™
Product Code. 11744346 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
Dry Wipe
Pre-moistened DI Wipes
Pre-moistened IPA Wipes
Saturation Solution:
100% pure deionized water
70% IPA/30% DI Water
Dry
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Saturation Solution Unit Size
11744346 Dry Wipe Dry Pack of 100
11734346 Pre-moistened DI Wipes 100% pure deionized water Pack of 100
12972780 Pre-moistened DI Wipes 100% pure deionized water Pack of 100
11754346 Pre-moistened IPA Wipes 70% IPA/30% DI Water Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11744346
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes.
Fisherbrand™ Clean-Wipes™ are ideal for cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes.
  • Cellulose and polyester blend cloth is soft and lint-free; will not scratch surfaces or fall apart like paper wipes
  • Reclosable container dispenses single wipes, eliminates contamination from dust and dirt
Order Info
Packed 100 wipes per polyethylene canister
Specifications
Product Type Dry Wipe
Length (English) 9 in.
Length (Metric) 22.8 cm
Width (English) 6 in.
Width (Metric) 15.2 cm
Material Polyester Cellulose
Dimensions (L x W) 9 x 6 in. (22.8 x 15.2 cm)
For Use With (Application) For cleaning or wiping optical parts, glassware, electrodes, cuvettes and microscopes.
For Use With (Equipment) For cleaning or wiping optical parts, Glassware, electrodes, cuvettes and microscopes
Saturation Solution Dry