Learn More
Fisherbrand™ Polypropylene Graduated Specimen Containers
Product Code. p-174404 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
133 mL
148 mL
237 mL
Capacity (English):
4.5 oz.
5 oz.
8 oz.
Sterility:
Non-sterile
Sterile
Closure Included:
No
Yes
Unit Size:
Pack of 100
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Capacity (English) Sterility Closure Included unitSize
11769378 133 mL 4.5 oz. Sterile No Pack of 500
11779378 148 mL 5 oz. Sterile Yes Pack of 100
11964395 133 mL 4.5 oz. Non-sterile Yes Pack of 500
11789378 237 mL 8 oz. Sterile No Pack of 500
11799378 237 mL 8 oz. Sterile Yes Pack of 100
11984375 237 mL 8 oz. Non-sterile No Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11769378
Only null left
Add to basket
Add to basket
Ideal for specimen collection, transportation and storage
  • Autoclavable
  • Graduated
  • Without lid
Order Info
Note: Sterile containers with lids are individually wrapped, and are sterile inside container. Nonsterile and sterile containers without lids are bulk packed.
Specifications
Closure Included No
Graduated Yes
Material Polypropylene
Sterility Sterile
Capacity (English) 4.5 oz.
Capacity (Metric) 133 mL