Learn More
FISHERBRAND™ FISHERBRAND™ Reusable Test Tubes with Rubber-Lined Screw Caps
Shop All Fisherbrand Traceable ProductsSpecifications
Specifications
| Certifications/Compliance | ASTM E982, Type VI, Class A |
| Closure Material | Phenolic |
| Capacity (Metric) | 50 mL |
| Length (Metric) | 150 mm |
| Material | Borosilicate Glass |
| Shape | Cylindrical |
| Label Type | Marking Spot |
| Liner Material | Rubber |
| Thread Size | 24-410 |
| Sterility | Non-sterile |
| Show More |