Learn More
Fisherbrand™ Hygrometer Replacement Battery
Product Code. p-33337 Shop All Fisherbrand Products
Change view
Click to view available options
Battery Type:
357
392
AAA
Product Type:
Button Cell Battery
Cylindrical Battery
Voltage:
1.5 V
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Type Product Type Voltage unitSize
15338754 357 Button Cell Battery 1.5 V Each
11735495 392 Button Cell Battery 1.5 V Each
15348754 AAA Cylindrical Battery 1.5 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15338754
Edge
Hygrometer Replacement Battery for use with Hygrometers.
  • Come with lithium, button cell and 'AAA' alkaline battery
  • The electrical requirements are 3 V, 1.5 V and 1.5 V respectively
Specifications
Product Type Button Cell Battery
Battery Type 357
Material Alkaline
Cell Type Button
For Use With (Equipment) For hygrometers
Temperature (English) Range -58°F to +302°F
Temperature (Metric) Range -50°C to +150°C
Voltage 1.5 V