Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
Product Code. 10136372 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Application):
1000mL erlenmeyer flask
250mL Erlenmeyer flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
10136372 250mL Erlenmeyer flask Each
10619474 50 mL Erlenmeyer Flask Each
10084622 500 mL Erlenmeyer Flask Each
10562963 1000mL erlenmeyer flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10136372
  • ONLINE EXCLUSIVE PRICE
    31.70€
    23.81€ / Pack of 1
    Save 7.89€
    24% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
  • Safe and secure attachment
  • Manufactured from strong and durable stainless steel
  • Provides superior stability
Specifications
Type Flask Clamp
For Use With (Equipment) Ultrasonic Cleaner
For Use With (Application) 250mL Erlenmeyer flask