Learn More
Fisherbrand™ Combs for Mini-Plus Wide Format Horizontal Gel Units, 1mm Thick
Product Code. 11563362 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
4
8
10
12
16
20
26
Volume (Metric) Sample:
22 μL
29 μL
40 μL
42 μL
52 μL
67 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample unitSize
11563362 16 29 μL Each
11543362 10 52 μL Each
11553362 12 40 μL Each
11573362 20 22 μL Each
11523362 4 42 μL Each
11533362 8 67 μL Each
11583362 26 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11563362
In Stock
Add to basket
Edge
Add to basket
For use with Mini-Plus Wide Format Horizontal Gel Units.
  • Recommended for high resolution gels and publication quality data
  • Supplied as standard
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Color White
For Use With (Equipment) Mini-Plus Wide Format Horizontal Gel Units
Type Comb
No. of Wells 16
Quantity 1 Each
Thickness (Metric) 1 mm
Volume (Metric) Sample 29 μL