Learn More
Fisherbrand™ Cleanroom Sticky Notes
Product Code. 15654583 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Dimensions (L x W):
3 x 3 in.
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15654583 3 x 3 in. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15654583
  • ONLINE EXCLUSIVE PRICE
    654.15€
    393.40€ / Pack of 24
    Save 260.75€
    39% Off
Estimated Shipment: 02-04-2027
Add to basket
Add to basket
Designed with low particulating cleanroom stock for easy cleanroom note-taking. Fisherbrand™ Sticky Notes are manufactured with 30# low particulating cleanroom stock and are 3 x 3 in. in size.
Designed with low particulating cleanroom stock for easy cleanroom note-taking. Fisherbrand™ Sticky Notes are manufactured with 30# low particulating cleanroom stock and are 3 x 3 in. in size.
  • Manufactured with 30# low particulating cleanroom stock
  • For use in cleanroom environments
  • 3 x 3 in.
  • 100 sheets per pad
Specifications
Material Low-particulate Stock
Width (English) Page 3 in.
Length (English) Page 3 in.
Description 100 sheets per page
Dimensions (L x W) 3 x 3 in.
Product Type Clean Notes