Learn More
Fisherbrand™ Cell Scrapers
Product Code. 11587692 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (English) Handle:
10 in.
15 in.
Length (English) Blade:
0.7 in.
1.18 in.
Size:
Small
Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Handle Length (English) Blade Size Unit Size
11587692 15 in. 1.18 in. Large Pack of 100
11597692 10 in. 0.7 in. Small Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11587692
In Stock
Add to basket
Edge
Add to basket
Easily harvest cells from tissue culture flasks
Fisherbrand Cell Scrapers are appropriate for a variety of cell culture vessels. The uniquely designed handle and blade reduce cell damage during the harvesting process.
  • Presterilized and individually wrapped in peel-open packaging
  • Nonpyrogenic
  • Polyethylene copolymer blade pivots to permit even contact with growth surface
  • Ribbed polystyrene handle provides secure grip and added control
  • Flexible joint between blade and handle eases entry into vessel
  • Pivoting blade improves position control to adjust for difficult angles during cell harvesting
Specifications
Length (English) Blade 1.18 in.
Length (Metric) Blade 3 cm
For Use With (Equipment) For a variety of cell culture vessels
Length (English) Handle 15 in.
Length (Metric) Handle 39 cm
Blade Material Polyethylene Copolymer
Product Type Cell Scraper
Size Large
Material Polystyrene (Handle)