Learn More
Fisherbrand™ Aluminum Foil, Standard-Gauge Roll
Product Code. 11749408 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (English):
25 ft.
197 ft.
492 ft.
Thickness:
18 μm
24 μm
Width (Metric):
30cm
30.5cm
46cm
Length (Metric):
7.6 m
60 m
150 m
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Thickness Width (Metric) Length (Metric) Unit Size
11749408 197 ft. 18 μm 30.5cm 60 m Each
11759408 25 ft. 18 μm 30cm 7.6 m Each
11769408 492 ft. 18 μm 30cm 150 m Each
11779408 25 ft. 24 μm 46cm 7.6 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11749408
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Edge
Add to basket
For sealing, weighing, and transferring samples
Ideal for weighing, sealing, and tranferring your samples.
  • Packed in individual dispenser box with cutter bar
  • Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Applications Sealing, Weighing, Transferring Samples
Length (English) 197 ft.
Product Type Foil
Width (English) 12 in.
Width (Metric) 30.5cm
Length (Metric) 60 m
Material Aluminum
Thickness 18 μm