Learn More
Fisherbrand™ Traceable™ Visual Alarm Timer
Product Code. 11719805 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Time Range:
Timing Capacity: 9999 min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Time Range Unit Size
11719805 Timing Capacity: 9999 min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11719805
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Includes
AAA batteries (2), Integrated magnet, Stand, Clip
Ideal for repetitive tests
Ideal for repetitive tests. Fisher Scientific™ Traceable™ Visual Alarm Timer has a switch which allows user to select either an audible alarm or bright flashing LED visual alarm.
  • Counts down from 9999 minutes in one-minute increments
  • Countdown resolution is 1 second for times under 99 minutes
  • Automatic memory resets the display to the last programmed time
  • Thru-zero feature allows counting down, alarming, then counting up
  • Digits are 1cm (3/8 in.) high and readable from 3M (10 ft.)
Specifications
Product Type Benchtop Timer
Time Range Timing Capacity: 9999 min.
Alarms Audible/Visible
Accuracy 0.0001
Resolution 1 second to 1 minute
Display LED
Battery Type AAA
Weight (English) 0.18 lb.
Weight (Metric) 0.08 kg
Includes Two AAA batteries, integrated magnet, stand, clip
For Use With (Application) For repetative tests
Certifications/Compliance NIST