Learn More
Fisherbrand™ Traceable™ Logger-Trac™ Temperature Datalogger Accessories
Product Code. 15338764 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Cable and Cradle
Waterproof Paper Sleeves
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15338764 Cable and Cradle Each
15308764 Waterproof Paper Sleeves Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15338764
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket
Accessories for Traceable™ Logger-Trac™ Temperature Datalogger.
15-081-145
  • Waterproof paper sleeves
  • For protecting temperature probe sensor
  • Transparent
15-081-148
  • USB standard A to Mini B, IC-9P or USB to serial converter
  • USB Version 2.0, ELV and RoHS compliant
  • Cable length: 1.5 m
  • Multiple cradles allow for batch programming up to 8 units
Specifications
Type Cable and Cradle
Certifications/Compliance RoHS compliant
Material Paper
For Use With (Application) Transfer recorded information in dataloggers to computer
For Use With (Equipment) Fisherbrand Traceable Logger-Trac Temperature Datalogger
Length (English) 5 ft.
Length (Metric) 1.5 m