Learn More
Honeywell™ Fireman Gloves
Click to view available options
Size: 10
Description
- Yellow water repellent cowhide grain leather glove
- Gunn cut, winged thumb, gauntlet type
- Hand lined with Kevlar®/cotton jersey
- Water repellent grain leather yoke at the base of thumb and middle finger
- Veins protection
- Split leather cuff
- Velcro™ strap
- Stitch with Kevlar™ thread
- Elasticated, with a fastening tab, the glove offers good support
- The silicone, water repellent cowhide grain leather ensures good flexibility at a low temperature, resisting water and offering good mechanical protection
- The Kevlar™ cotton lining provides protection against cuts and thermal hazards
- The Kevlar™ thread assembly ensures sturdy resistance of the seams with regard to the heat and wear, while offering the best possible thermal insulation
Specifications
Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 407-2004 Thermal hazards |
Color | Yellow |
Material | Cowhide Leather, Cotton |
Size | 10 |
Thickness (Metric) | 1.1 to 1.3 mm |