Learn More
Thermo Scientific™ Finnpipette™ Focus Fixed Volume Pipettes
Thermo Scientific™ Finnpipette™ Focus Short Fixed Volume Pipette has been discontinued, and is no longer available.
Brand: Thermo Scientific™ 4601080
Additional Details : Weight : 1.00000kg
Description
Thermo Scientific™ Finnpipette™ Focus™ Short Fixed Volume Pipette has been discontinued, and is no longer available. Please take a moment to learn about our current available manual pipetting systems.
Fixed-Volume Models
- 1 to 25μL models feature “super blow-out” new streamlined design
- Constructed of tough, lightweight polymers; fully autoclavable for absolute sterility
- Easily disassembled for cleaning
Compatible with:
For use with Thermo Scientific Finntip tips. Also compatible with all Fisherbrand™ Universal-Fit tips, Fisherbrand SureOne, Fisherbrand Ergo-F, Large Orifice, Large Orifice/Genomic Aerosol Barrier, and Low-Retention tips.
Specifications
±0.4% | |
Autoclavable | |
Polymer | |
Fixed Volume Pipette | |
0.3% |
Finnpipette Focus Short Fixed, Volume: 200µL, Discontinued | |
Thermo Scientific™ FinnTip™ Tips | |
Sterile | |
200 μL |