Learn More
Polyco Healthline Fine Touch 1-ply White C Fold Hand Towels
Product Code. 30130274
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 2640
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30130274 White Pack of 2640
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30130274 Supplier Polyco Healthline Supplier No. PHTC1W
  • ONLINE EXCLUSIVE PRICE
    107.00€
    40.34€ / Pack of 2640
    Save 66.66€
    62% Off
View all promotions
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
The Fine Touch 1 ply C fold towels are self presenting, ensuring a hygienic and a wasteless dispensing option.
  • 1 ply
  • C-Fold interleaved for efficient dispensing with minimum waste
  • Disposable product for easy and quick handling, increased hygiene and reduced risk of contamination
  • Embossed finish ensuring softness and great comfort
  • Available in Green and Blue
Specifications
Product Type Hand Towel
Length (English) 12.20 in.
Length (Metric) 310 mm
Width (English) 8.54 in.
Width (Metric) 217 mm
Material Paper
Autoclavable Not Autoclavable
Color White
Disposable Single-use
Ply 1-ply