Learn More
Macherey-Nagel™ MN 615 Grade Round Format Cellulose Qualitative Filter Paper
Use for a wide range of general laboratory filtrations. MN 615 Grade Round Format Cellulose Qualitative Filter Paper provides medium fast filtration.
Brand: Macherey-Nagel™ 431012
Additional Details : Weight : 1.00000kg
Description
- For general purpose filtrations
- Qualitative filter papers are manufactured from the same raw materials as the ashless grades and are particularly suited for general laboratory filtrations
- The average ash content is about 0.1%
- The amount of a-cellulose is about 95%.
- Medium fast filtration
Specifications
Disc | |
Medium fast | |
MN 615 | |
0.16 mm | |
125 mm |
Cellulose | |
Qualitative analysis | |
100 Pack | |
Filter Paper |