Learn More
Agilent BioTek Filters for ELx800 Absorbance Microplate Reader
Accessory for use with ELx800 Absorbance Microplate Reader.
Brand: Agilent BioTek 7334660
Additional Details : Weight : 0.80000kg
Description
Accessory for use with ELx800 Absorbance Microplate Reader.Specifications
Filter | |
ELx800 Absorbance Microplate Readers | |
660 nm |