Learn More
Sartorius™ Filter Weighing Pan
For use with Secura™ and Quintix™ lab balances.
Brand: Sartorius™ YFW01SQP
Additional Details : Weight : 0.90718kg
Description
- 130 mm diameter
- Accessory item
- Conversion kit for 0.01 mg balances included
Specifications
Secura and Quintix Balance | |
Weighing | |
130 mm diameter | |
Weighing Pan |