Learn More
Witeg™ Filter Funnels
Witeg™ Filter Funnels include a filter disc for separating materials from liquid solutions.
Brand:  Witeg™ 2 367 903
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11827820
  • 159.00€ / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Borosilicate glass construction
  • Variety of volumetric capacities available
  • Variety of disc porosities available
  • Variety of stem sizes available
Specifications
Specifications
Filter funnel
1000 mL
120mm
120mm
22mm
3