Learn More
Lenz Laborglasintrumente Filter Funnels
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07195279
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10527851
  • 784.40€ / Each
Estimated Shipment: 17-06-2024
Description
  • Filter funnels
  • Material: borosilicate glass 3.3
Specifications
Specifications
4 L
400 mm
2
Borosilicate Glass 3.3
175 mm