Learn More
UVP Filter excitation for automated BioLite 533nm to 587nm UVP
Brand:  UVP 38-0341-04
Additional Details : Weight : 0.20000kg
Product Code. 12843079
  • 670.00€ / Each
Estimated Shipment: 02-08-2024