Learn More
Duran™ Filter Candle With Tube
The Filter Candle with Tube is suitable for the extraction of supernatant solutions and also for the separation of solids from a carrier gas. Available with porosities ranging from 1 to 4.
Brand:  Duran™ 258505406
Additional Details : Weight : 0.08700kg
 View more versions of this product
Product Code. 10121741
  • 76.25€ / Each
Estimated Shipment: 06-12-2023
Description
  • Clear DURAN™ borosilicate glass 3.3
  • Conical filter
  • With tube
Specifications
Specifications
Borosilicate Glass 3.3
4
24 x 65 mm (Filter)
20 mm (Tube)
10 to 16 μm
1 Ea.
200 mm (Tube)