Learn More
Lenz Laborglasintrumente Filter Adapters
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 07189027
This item is not returnable.
View return policy
Description
- Filter adapters
- For the insertion of filter crucibles
- Material: borosilicate glass 3.3
Specifications
Borosilicate Glass (3.3) | |
110 mm | |
27 mm |