Learn More
Thermo Scientific™ BspLI (NlaIV) (10 U/μL)
Cut at GGN^NCC sites with BspLI (NlaIV) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BmiI, NlaIV, PspN4I).
94.00€ - 382.00€
Specifications
Concentration 10 U/μL
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10898990
View Documents Promotion Details
Thermo Scientific™
ER1151
200 U
94.00€
200 units
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10314720
View Documents Promotion Details
Thermo Scientific™
ER1152
1000 U
382.00€
1000 units
View alternative products
Save Up to 
»
In Stock
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications
Specifications
10 U/μL