Description
- Cole-Parmer offers a variety of barbed luer adapters to meet your application's needs.
- Standard hose barb for use with flexible tubing.
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination.
- Wide-bore hose barb for enhanced fluid path flow.
- Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene.
- Manufactured in compliance with ISO 594-1 standards, these fittings are inspected to be free from contaminants, and are fully traceable.
- All parts meet tight dimensional tolerances for increased reliability and process purity.
- Choose from a range of sizes, 1⁄16 to 1⁄4in. with a variety of connection options.
- Sold in packs of 25.
Specifications
Specifications
| Material | Polypropylene |
| Packaging Quantity | 25/Pk. |
| Product Type | Female Luer Fitting |