Learn More
Lenz Laborglasintrumente Extractors Acc. to Thielepape
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05370049
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 10306221
  • 508.80€ / Each
Estimated Shipment: 20-08-2024
Description
  • Extractor head with extraction thimbles (porosity: 0)
  • Made of DURAN™ tubing
  • Dimroth condenser with thread GL 14
  • Round bottom flask
Specifications
Specifications
500 mL
0
NS 45/40