Learn More
SI Analytics™ Exchange Unit
SI Analytics™ Exchange Unit is a unit that includes an integrated chip for reagent dat, tubes, a titration tip, and more.
800.00€ - 820.00€
Specifications
Color | Brown glass bottle for titrant |
---|---|
Container Size | GL 45 |
Type | Exchange Unit |
Includes | Integrated chip for reagent data |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11358310
|
SI Analytics™
285220320 |
20 mL |
800.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
13476979
|
SI Analytics™
285220300 |
5 mL |
820.00€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
- Includes integrated chip for reagent data
- Comes with a brown glass bottle for titrant
- GL 45
- S 40 bottle adapter
- Includes tubes, drip tubes, and titration tip
Specifications
Brown glass bottle for titrant | |
GL 45 | |
Exchange Unit | |
Integrated chip for reagent data |