Specifications
| Product Type | ES Type 1 Tag Out |
| Alternate Part Number | ES-TYPE 1-145X85 SAFETY TAGS FR |
| Color | Black/Yellow on White |
| Material | Polyester |
| For Use With (Application) | Safety Identification |
| Height (English) | 5.71 in. |
| Height (Metric) | 145 mm |
| Quantity | 10 Pk. |
| Temperature Range (English) | -40°F to +212°F |
| Temperature Range (Metric) | -40°C to +100°C |
| Show More |