Learn More
Lenz Laborglasintrumente Erlenmeyer-Flask Duran
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03022452
Additional Details : Weight : 0.10000kg
Product Code. 10524001
  • 189.74€ / Pack of 10
Estimated Shipment: 04-07-2024
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume /mL)
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
300 mL
NS 24/29
87 mm