Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit
3M Versaflo™ TR-600 PAPR Accessory, Battery Charger Kit is used with 3M™ Versaflo™ TR-600 series air purifying device.
Brand: 3M™ 7000144504
212.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.72000kg Transport : UN number : 3481 Chem class : 9
Description
- Charges TR-630 standard battery or TR-632 high capacity battery in less than 4 hours
- Batteries may be stored on the charger, fully charged, ready for immediate use
Specifications
Single Station Battery Charger Kit | |
3M Versaflo Powered Air Turbo TR-600 Series | |
TR-640 Cradle, Power Supply and Lead | |
TR-641E |