Learn More
Olympus™ Ergonomic Eyepiece Observation Tubes
Reduce strain by using ergonomically designed observation tubes. Olympus™ Ergonomic Eyepiece Observation Tubes permit a more neutral erect working posture by using adjustable, longer optical path eyepiece tubes.
Brand: Olympus™ K10017509
Additional Details : Weight : 0.01000kg
Description
- Increased eyepiece observation tube length.
- Adjustable format.
- Ergonomically designed.
Specifications
Eyepiece Tubes | |
BX and CX Microscopes |