Learn More
Invitrogen™ Eotaxin Mouse ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand:  Invitrogen™ 900-K68
Additional Details : Weight : 0.03500kg
Product Code. 17819055
  • 330.00€ / Pack of 10
Estimated Shipment: 18-06-2024
Description
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
Specifications
P48298
20292
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
ELISA
CCL11