Learn More
3M™ Emphaze™ Polyimide Film Tape 5413
Product Code. 17135034 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
36 yd. x 0.5 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17135034 36 yd. x 0.5 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17135034 Supplier 3M™ Supplier No. 7100010443
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
Made with polyimide film and a silicone adhesive, useful for PCB solder masking and other high temperature applications.
Use 3M™ Polyimide Film Tape 5413 for PCB solder masking and other high temperature applications within a range of 73 to 260°C. This amber colored, 0.07 mm thick tape is made with a polyimide film and a silicone adhesive. It also features a polyethylene tape core instead of cardboard.
  • Silicone adhesive's high temperature performance reduces adhesive transfer, helping to eliminate cleaning
  • Polyimide film provides excellent release, as it doesn't soften and stays dimensionally stable at high temperatures
  • Surfaces are protected due to the tape's flame retardancy, and chemical and radiation resistance
TRUSTED_SUSTAINABILITY
Specifications
Adhesive Type Silicone
Color Amber
Material Polyimide
Dimensions (L x W) 36 yd. x 0.5 in.
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 0.5 in.
Width (Metric) 1.2 cm