Learn More
Thermo Scientific™ Emission filters for Thermo Scientific Varioskan™ LUX Multimode Microplate Readers
Varioskan LUX multimode microplate reader can be equipped with a range of measurement technologies including absorbance, fluorescence intensity, luminescence, AlphaScreen and time-resolved fluorescence. Additional emission filters expand the instrument functionality and application flexibility.
780.00€ - 912.00€
Products 13
Product Code Brand Product Type Wavelength For Use With (Application) Price Quantity & Availability  
Product Code Brand Product Type Wavelength For Use With (Application) Price Quantity & Availability  
12449757
View Documents Promotion Details
Thermo Scientific™
F48020
Emission Filter 480 nm BRET donor using wtRluc
912.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15360787
View Documents Promotion Details
Thermo Scientific™
F49210
Emission Filter 492 nm General Terbium measurement
792.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
12459757
View Documents Promotion Details
Thermo Scientific™
F51060
Emission Filter 510 nm Chroma-glo luciferase CBG68 or CBG99 assay
912.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
12469757
View Documents Promotion Details
Thermo Scientific™
F51530
Emission Filter 515 nm BRET2 acceptor using GFP2
912.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15370787
View Documents Promotion Details
Thermo Scientific™
F52010
Emission Filter 520 nm LanthaScreen measurement of Tb acceptor; HTRF donor measurement of TbGreen
792.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
17128905
View Documents Promotion Details
Thermo Scientific™
F52530
Emission Filter 525 nm Pierce green Renilla luciferase assay
912.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
12479757
View Documents Promotion Details
Thermo Scientific™
F53030
Emission Filter 530 nm BRET acceptor using eYFP
912.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15300797
View Documents Promotion Details
Thermo Scientific™
F57177
Emission Filter 571 nm AlphaScreen measurement
794.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
12489757
View Documents Promotion Details
Thermo Scientific™
F61060
Emission Filter 610 nm Chroma-glo luciferase CBR assay
910.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15320797
View Documents Promotion Details
Thermo Scientific™
F61520
Emission Filter 615 nm AlphaLisa measurement
794.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15310797
View Documents Promotion Details
Thermo Scientific™
F62010
Emission Filter 620 nm General Europium measurement; LanthaScreen measurement of Eu donor; HTRF acceptor measurement of TbGreen; HTRF donor measurement of EuRed and TbRed
780.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
17138905
View Documents Promotion Details
Thermo Scientific™
F640LP
Emission Filter 640 nm Pierce red firefly luciferase assay
912.00€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
15380787
View Documents Promotion Details
Thermo Scientific™
F66510
Emission Filter 665 nm LanthaScreen measurement of Eu acceptor; HTRF acceptor measurement of EuRed and TbRed
782.00€
Each
View alternative products
Save Up to 
»
In Stock
Description
Varioskan LUX comes equipped with a range of measurement technologies including absorbance and fluorescence intensity with optional luminescence, AlphaScreen and TRF. The instrument selects the measurement wavelength using monochromators or optical filters, depending on which is optimal for each measurement technology. A wide range of optional filters extends the application area remarkably, including luciferase assays, Bioluminescence Resonance Energy Transfer (BRET) assays, Homogeneous Time Resolved Fluorescence (HTRF) assays and many other.
  • Extensive range of different type of emission filters are available for various applications when performing luminometric, TRF or Alpha technology assays, such as TR-FRET, hTRF, BRET, NanoBRET, AlphaScreen/AlphaLisa and all multiplexing assays using these detection technologies.
  • Excitation filters for TRF and AlphaScreen/AlphaLisa measurements are permanently fixed inside the corresponding detection technology optical module.
  • Emission filter installation is effortless. The instrument filter wheel is easy to access, and the correct parameters are quickly set in simple workflow in Thermo Scientific SkanIt software. SkanIt software controlled installation process also prevents accidental mistakes during the process and therefore ensures proper operation.
  • In most luminescence measurements, there is only one source of light in the system and therefore the most efficient detection is received by collecting all the light emitted by the sample, independent on the wavelength. Therefore, these assays do not require any wavelength selection. However, certain in applications, like BRET or any luminometric multiplexing assay, where multiple light sources are present in the sample, filters are used to collect the light produced at specific wavelength.
  • All Varioskan LUX filters can be used for any filter based detection technology (luminescence, TRF or Alpha technology), depending on the application. Please contact technical support for more information concerning your specific application requirements.