Learn More
Merck Millipore Petri Dishes and Petri-Pad™ Dishes
Available with or without absorbent pads
526.00€
Requires: When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 045 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)
Products 1
Product Code Brand dimType Quantity Price Quantity & Availability  
Product Code Brand dimType Quantity Price Quantity & Availability  
11993856
View Documents
Merck Millipore
PD20047S5
ONLINE EXCLUSIVE PRICE
562.00€
526.00€ / Pack of 600
Save 36.00€
6% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 06-02-2026
Add to basket
Description
Description
Available with or without absorbent pads
Petri Dishes
  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose