Learn More
Fisherbrand™ Plain Economy PTFE Stir Bar
Product Code. 11597572 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
12 mm
25 mm
40 mm
50 mm
Diameter (Metric):
3 mm
8 mm
Unit Size:
Each
Pack of 10
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
11597572 25 mm 8 mm Pack of 10
11785134 25 mm 8 mm Pack of 100
11517582 50 mm 8 mm Pack of 10
11587572 12 mm 3 mm Pack of 10
11775134 12 mm 3 mm Pack of 100
11507582 40 mm 8 mm Pack of 10
10373692 40 mm 8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11597572
In Stock
Add to basket
Edge
Add to basket
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 8 mm
Length (Metric) 25 mm
Quantity 10/Pk.